Drucken

Anti-HSPA5 Goat Polyclonal Antibody

Lieferant: Everest Biotech
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

EB12856
EVBIEB12856EA 339 EUR
EVBIEB12856
Anti-HSPA5 Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 75kDa band observed in lysates of cell lines HepG2, HeLa and MCF7 (calculated MW of 72.3kDa according to NP_005338.1). Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: HSPA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR