Restriction enzymes, HindIII, FastDigest™, Fermentas
Lieferant: FERMENTAS
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
FD0504
FD0505
FD0504EA
53.25
EUR
FD0504
FD0505
Restriction enzymes, HindIII, FastDigest™, Fermentas
Nukleinsäure-Reagenzien
Restriktionsenzym
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.