Drucken
Restriction enzymes, HindIII, FastDigest™, Fermentas
Lieferant: FERMENTAS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
FD0504 FD0505
FD0504EA 53.25 EUR
FD0504 FD0505
Restriction enzymes, HindIII, FastDigest™, Fermentas
Nukleinsäure-Reagenzien Restriktionsenzym
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.