Drucken
Restriction enzymes, NotI, FastDigest™, fermentas
Lieferant: FERMENTAS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
FD0594 FD0595
FD0594EA 89 EUR
FD0594 FD0595
Restriction enzymes, NotI, FastDigest™, fermentas
Nukleinsäure-Reagenzien Restriktionsenzym
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Designed to complete digestion in 5 to 15 minutes
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications