Glucose (GO) assay kit, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
GAGO20-1KT
GAGO20-1KTEA 130 EUR InStock
GAGO20-1KT
Glucose (GO) assay kit, Supelco®
Test-Systeme für Lebensmittel und Getränke
This kit is for the quantitative, enzymatic determination of glucose in food and other materials.
Glucose is oxidised to gluconic acid and hydrogen peroxide by glucose oxidase. Hydrogen peroxide reacts with o-dianisidine in the presence of peroxidase to form a coloured product. Oxidised o-dianisidine reacts with sulfuric acid to form a more stable coloured product.
The intensity of the pink colour measured at 540 nm is proportional to the original glucose concentration.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge