Drucken
Ceran® hotplates, EB series
Lieferant: GESTIGKEIT HARRY
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
11EB 22EB
GEST11EBEA 0 EUR
GEST11EB GEST22EB
Ceran® hotplates, EB series
Heizgeräte Heizplatten
Glass ceramic heating plate, which is extremely resistant to breakage and changes of temperature, will not distort, is permeable to ultra-violet light and also highly acid-resistant.
  • Uniform heating across full surface
  • Insulated stainless steel housing
  • Temperature control by an NiCr-Ni sensor (regulation from 50 to 500 °C)
In the EB Series the electrical connection between the controller and the hotplate is protected by a flexible metal hose.