Drucken
Heat exchanger and circulator packages
Lieferant: HAAKE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
222-1937
HAAK222-1937EA 0 EUR
HAAK222-1937
Heat exchanger and circulator packages
Umwälzthermostate Geschlossene Umwälzsysteme Wärmeumwälzthermostate
SC100- A10 refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −10 to +100°C and maximum bath volume of 6 litres.
  • Powerful bath circulator with a stainless steel reservoir
Lieferumfang: Circulator package includes SC100-A10 and 3 m silicon hose