Drucken
Conductivity sensor, inductive, 3700 SC
Lieferant: Hach
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
D3708E2T.99
HACHD3708E2T.99EA 1962 EUR
HACHD3708E2T.99
Conductivity sensor, inductive, 3700 SC
Elektroden Leitfähigkeitselektroden
The innovative technology of Hach’s inductive conductivity sensors eliminates polarization and electrode coating problems in harsh environments. The sensor comes complete with standard sensor cable length of 6 m, digital gateway, and a 1 m digital extension cable.
  • Sanitary (CIP) flange style: 2-inch flange, special cap, and EPDM compound gasket
  • Conforms to provisions of 3-A Sanitary Standards