Drucken
Permanent marker set, dual-tip, Sharpie®
Lieferant: HEATHROW
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
HS15094
HEATHS15094EA 68.1 EUR
HEATHS15094
Permanent marker set, dual-tip, Sharpie®
Schreibstifte Permanentmarker
Have the convenience of a fine tip and an ultra fine tip all in one marker. Brightly colored 8-piece set is ideal for research, industrial, biomedical and educational laboratories. When used on glass, metal or porcelain surfaces, ink will not smear when subjected to water. Wash off ink with solvents for repeated use.
  • Specially-formulated, water-resistant ink
  • Assorted colours: Berry; Black; Lime; Navy;Orange; Purple; Sky Blue; Turquoise
  • Fine point tip is compatible with the eCount™