Drucken
Assistent® Stirring Spatula
Lieferant: GLASWARENFABRIK KARL HECHT
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
40734010
HECH40734010EA 7.5 EUR
HECH40734010
Assistent® Stirring Spatula
Spatel
Designed for several uses, such as mixing samples and solutions in test tubes, resolution of substrate, additions of small reagent volumes, or releasing blood clots from the side of test tube walls.
  • Stirring rod for a wide variety of laboratory applications
Made from polystyrene, crystal clear.