Drucken
Condensers, Allihn, for Soxleth extractors
Lieferant: GLASWARENFABRIK KARL HECHT
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
41153045
HECH41153045EA 73 EUR
HECH41153045
Condensers, Allihn, for Soxleth extractors
Kühler
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With NS cone. Water connection with glass nozzles.