Drucken

Elan 6000/5000 plasma setup solution Hg for ICP

Lieferant: HONEYWELL
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

E2185-100ML
HONCE2185-100MLEA 785.38 EUR
HONCE2185-100ML
Elan 6000/5000 plasma setup solution Hg for ICP
Standards Kationischer Standards Mehrelement-Standards

Certified Reference Material (CRM).


10 g/L each of Ba, Cd, Ce, Cu, Ge, Pb, Mg, Rh, Sc, Tb, Tl in HNO₃ 2%

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR