Drucken
HPLC Columns, μPAC™ Neo
Lieferant: Thermo Fisher Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
COL-NANO110NEO
HYPECOL-NANO110NEOEA 0 EUR
HYPECOL-NANO110NEO
HPLC Columns, μPAC™ Neo
Chromatographiesäulen
μPAC Neo column line offers higher resolution ensuring that you can extract the maximum amount of information from highly complex samples using low-flow chromatography.
  • Operating at lower back pressures extends column lifetime
  • Designed with a perfect flow path order, eliminating heterogeneous flow paths
The μPAC Neo column range is the ideal choice for a variety of proteomics experiments, including fast label-free quantitation, comprehensive identification, and single cell analysis.
These columns operate at moderate pressures, allowing for longer flow paths that increase separation performance and sensitivity.
The unique micromachining manufacturing of μPAC Neo column line produces columns that are virtually identical. This results in increased injection-to-injection and column-to-column reproducibility, providing greater confidence in the results throughout the duration of a research study.
The ordered flow path of the columns minimises dispersion to the overall separation, resulting in sharper and more intense chromatographic peaks.