Drucken
EASY-Spray™ PepMap Neo UHPLC columns
Lieferant: Thermo Fisher Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
ES75150PN
HYPEES75150PNEA 0 EUR
HYPEES75150PN
EASY-Spray™ PepMap Neo UHPLC columns
Chromatographiesäulen
The Thermo Scientific EASY-Spray PepMap Neo UHPLC columns are perfect for bottom-up proteomics. Packed at higher pressure and rated to 1500 bar, they provide consistent column-to-column performance, long column lifetime, and excellent efficiency. These benefits are true at any pressure.
  • Sample amount is limited
  • Analytical UHPLC does not provide sufficient sensitivity
  • Workflow simplicity is key
  • High sensitivity is required to identify proteins and peptides at low expression levels
  • Analyses are done in a targeted and untargeted way for screening and verification