Drucken

Digitonin, weißes Pulver

Lieferant: MP BIOMEDICALS
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Gefahr

0215948082 0215948090
ICNA0215948082EA 173 EUR
ICNA0215948082 ICNA0215948090
Digitonin, weißes Pulver
Digitonin

Digitonin is a mild nonionic detergent. It permeabilizes plasma membranes of eukaryotic cells by complexing with membrane cholesterol and other unconjugated β-hydroxysterols.


  • Water soluble
  • Critical Micelle Concentration (CMC): <0.5 mM (20…25 °C)(Lit.)
  • [α]20/D −54 °, c = 2.8 in methanol (Lit.)


Digitonin has used for cholesterol determination, use of digitonin-permeabilised adipocytes for cAMP studies, solubilisation of membrane proteins. The water soluble form can be used for the isolation of mitotic chromosomes. Mild nonionic detergent used to solubilise receptors and permeabilise cellular and nuclear membranes.


Soluble in boiling water (1g/10 ml). Heat for 15 minutes at 98 °C. Cool and place at 4 °C for 3 hrs and filter off any precipitate. Soluble in absolute alcohol (1 g / 57 ml) and 95% alcohol (1 g).

Formel: C₅₆H₉₂O₂₉
MW: 1229.312 g/mol
Melting Pt: 230…240 °C (Zers., Lit.)
MDL Number: MFCD00077729
CAS-Nummer: 11024-24-1
EINECS: 234-255-6
UN: 3462
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR