Drucken

Lecithin aus Eigelb ≥96%, Pulver

Lieferant: MP BIOMEDICALS
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

02160059.2 02160059.5 0216005905 02160059.1 0216005901
ICNA02160059.2EA 152 EUR
ICNA02160059.2 ICNA02160059.5 ICNA0216005905 ICNA02160059.1 ICNA0216005901
Lecithin aus Eigelb ≥96%, Pulver
Lecithin aus Eigelb

Lecithin is the major membrane phospholipid in eukaryotic cells. It also serves as a reservoir for several lipid messengers.


Lecithin is used for preparation of vesicle suspensions commonly called liposomes or as monolayers. It is the source of the bioactive lipids lysophosphatidylcholine, phosphatidic acid, diacylglycerol, lysophosphatidylcholine, plateletactivating factor, and arachidonic acid.


A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.


Soluble in fat, paraffin, ether, ethanol, chlorinated hydrocarbons(100 mg/ml, CHCl3). Dispersible in water.

Storage Temperature: Kühlschrank
MDL Number: MFCD00131449
CAS-Nummer: 93685-90-6
EINECS: 297-639-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR