Drucken

L-Methionin ≥99%, weißes kristallines Pulver Reagenz für die Zellkultur

Lieferant: MP BIOMEDICALS
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

0219470791 0219470725 0219470780 0219470790
ICNA0219470791EA 886 EUR
ICNA0219470791 ICNA0219470725 ICNA0219470780 ICNA0219470790
L-Methionin ≥99%, weißes kristallines Pulver Reagenz für die Zellkultur
L-Methionin

Storage: Store at Room Temperature (15-30 °C).
Methionine is one of the common sulfur-containing amino acids. Methionine is a common methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol, and choline.


  • Cell Culture Reagent Grade
  • Presentation: White Crystalline Powder
  • pKa: 2.28, 9.21 (Lit.)
  • Soluble at 2.5 g to 100 mL of water.

Formel: C₅H₁₁NO₂S
MW: 149.21 g/mol
Siedepunkt: 278
Melting Pt: 280…282 °C (Lit.)
Dichte: 1.373 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00063097
CAS-Nummer: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004

Order Now

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
pH (1% aq soln) 5.6-6.1
Loss on Drying ≤0.3%
Optical Rotation (c=2, 6N HCl) +22.4° to 24.7°
Residue on Ignition ≤0.4%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR