Drucken
LB Agar, Lennox Large Capsules, MP Biomedicals
Lieferant: MP BIOMEDICALS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
113002326
ICNA113002326EA 270 EUR
ICNA113002326
LB Agar, Lennox Large Capsules, MP Biomedicals
Zellkulturmedien
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
  • Proprietary media for enhanced cell yield
  • Convenient packaging formats for a variety of applications and budgets
  • Proven and reliable reputation
  • High-quality raw materials and additives
  • Encapsulated media
  • No weighing, dust and cleanup
  • Drop capsules into water and autoclave
LB Lennox Agar capsules is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. The ultimate in ease and convenience, LB Lennox Agar in a capsule form eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder. The growth characteristics are the same as LB Lennox Agar powder. LB Lennox Agar Medium supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesize. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
Zertifizierungen: Molecular Biology Certified™.