Drucken
Buffer, SSC 20X powder for molecular biology, MP Biomedicals
Lieferant: MP BIOMEDICALS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
11RIST2050
ICNA11RIST2050EA 99 EUR
ICNA11RIST2050
Buffer, SSC 20X powder for molecular biology, MP Biomedicals
Puffer Elektrophorese-Puffer Hybridisierungspuffer
SSC buffer used in electrophoresis and in sedimentation studies of DNA-histone aggregates. SSC is used as a hybridisation buffer in immunoblotting and DNA microarray protocols.