Drucken

Anti-IgG F(c) Rabbit Polyclonal Antibody

Lieferant: ImmunoReagents
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

IR2490
IMMRIR2490EA 169 EUR
IMMRIR2490
Anti-IgG F(c) Rabbit Polyclonal Antibody
Antikörper
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Zertifizierungen: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Bestellinformation: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR