Drucken

Polyethylenglycol 6.000, Schuppen, ultrarein

Lieferant: ALFA AESAR
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

J19972.A1 J19972.A7
J19972.A1EA 80.2 EUR
J19972.A1 J19972.A7
Polyethylenglycol 6.000, Schuppen, ultrarein
Polyethylenglycol 6.000

Application includes differential precipitation of DNA, enhances hybridisation rate of nucleic acids, improves efficiency of end-labeling with T4 polynucleotide kinase, used in ligation of blunt ended DNA, precipitates bacteriophage ± particles and plasmid DNA, used in preparing single-stranded M13 DNA and promotes cell fusion.

Formel: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Melting Pt: 60…63 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD01779614
CAS-Nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,07568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR