Destaining bags, ethidium bromide
Lieferant: ALFA AESARGesamtbewertung: 0
Bewertung: von 5
Ethidium bromide destaining bags are used to safely remove ethidium bromide from nucleotide solutions and gels during overnight treatment. Each bag extracts up to 5 mg of Ethidium Bromide from solution, thus minimising the exposure of research personnel to this material.
- The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration
- Provides a safer, more economical means for disposal of hazardous waste
- Also removes Coomassie stains
Applications: For removal of ethidium bromide from nucleotide solutions and gels. Maximum capacity is 5mg ethidium bromide. Also removes Coomassie stains