Drucken
Histology baths
Lieferant: KLINIPATH
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
205200
KLIN205200EA 1210 EUR
KLIN205200
Histology baths
Bäder Wasserbäder, ohne Umwälzung
Compact histology baths designed to efficiently remove wrinkles and distortions in parafin wax sections.
  • Round or square baths with light
  • LED light provides excellent visibility and contrast
  • Digital touch control
  • Warm areas for drying slides, space forup to 18 (square) or 27 slides (round bath)