Drucken
Osmometer, semi-micro, K-7400S
Lieferant: KNAUER
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
A0006AC
KNAUA0006ACEA 7610 EUR
KNAUA0006AC
Osmometer, semi-micro, K-7400S
Klinisches Diagnosezubehör Klinische Analysegeräte
The K-7400S semi-mirco osmometer enables the measurement of osmolality as well as the freezing point of aqueous solutions. The proven technology of freezing point determination in combination with the robust and intelligent design of the device allows fast and reproducible measurements.
  • Two-point or optional three-point calibration
  • LED display of two rows with 24 characters
The instrument is equipped with a peltier cooler and an integrated microprocessor controlling the automated measurement. The freezing point osmometer is a standalone device that optionally can be equipped with a printer. Furthermore, the device can be controlled via the EuroOsmo 7400 software.
Zubehörinformationen: EuroOsmo® 7400 is a user-friendly software developed for the freezing point osmometers K-7400 and K-7400S. It allows control of the instruments and recording of the measured data. The software automatically plots the temperature curve for each measurement and calibration and allows saving of the measured values. As with the instrument itself, the user can choose between freezing point depression or osmolality to be displayed also in the software. The data can optionally be exported into xls or txt files for further analysis. For archival storage a detailed analysis report can additionally be printed.