ThermostatCare Heat Transfer Liquids for Circulating Thermostats
Lieferant: KRUESSGesamtbewertung: 0
Bewertung: von 5
Heat transfer liquid for circulating thermostats tailored for use with PT31/PT80.
ThermostatCare was developed by A.Krüss Optronic to serve as a supreme heat transfer liquid which protects thermostats against corrosion and biofouling even for prolonged operation times.
Additionaly a version including an coloured indicator that indicates a degredation of stability is also offered.