Drucken
Clipped corner microscope slide, Snowcoat®
Lieferant: LEICA MICROSYSTEMS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
3800381
LEIC3800381EA 0 EUR
LEIC3800381
Clipped corner microscope slide, Snowcoat®
Objektträger Mikroskop-Objektträger
Snowcoat® Clipped Corder Slides are clipped at 45° angles to help reduce glass breakage and fragmentation when loaded into slide printers.
  • Available in a variety of colours
  • Specialty painted surface
  • Efficient scanning
Specialty painted surface formulated to optimise Ink Jet printing and bar code scanning efficiency. These slides have been optimised for use with the Leica printer.