Drucken
Biology prepared microscope slides, school set B
Lieferant: LIEDER JOHANNES
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
602C 605C 620E 621D 624D 625D 627C 630C 634D 635D 637D 646D 647C 649C 600 601D 606D 607D 610D 611B 615D 618E 619C 622G 626C 631C 636E 640C 642C 643C 644C 645D 648C 650E
LIED602CEA 8.6 EUR
LIED602C LIED605C LIED620E LIED621D LIED624D LIED625D LIED627C LIED630C LIED634D LIED635D LIED637D LIED646D LIED647C LIED649C LIED600 LIED601D LIED606D LIED607D LIED610D LIED611B LIED615D LIED618E LIED619C LIED622G LIED626C LIED631C LIED636E LIED640C LIED642C LIED643C LIED644C LIED645D LIED648C LIED650E
Biology prepared microscope slides, school set B
Objektträger Dauerpräparate
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
Lieferumfang: Supplied on high quality ground edge slides (26×76 mm).