Drucken
Parasitology prepared slides
Lieferant: LIEDER JOHANNES
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
74900
LIED74900EA 338.7 EUR
LIED74900
Parasitology prepared slides
Objektträger Dauerpräparate
Blood smear (t.s) tranverse section (w.m) whole mount (l.s) longitudinal section.
  • Sleeping disease (sm)
  • Malaria in rodents (sm) with vegetative forms and schizogony stages
  • Section of muscle showing the parasites in Miescher’s tubes
  • Honey bee dysentery (t.s) of diseased bee intestine
  • Causes coocidiosis in rabbit liver (t.s) shows parasites in all stages
  • Beef liver fluke (w.m) of adult flat mount and carefully stained
  • Fasciola hepatica, ova (w.m)
  • Taenia spec. or Moniezia spec., tapeworm, scolex (head) (w.m)
  • Dog tapeworm, mature proglottids (w.m).
  • Tapeworm, proglottids in different stages (t.s)
  • Dwarf tapeworm, proglottids (w.m)
  • Cyst wall and scolices sec.
  • Roundworm of human, adult female (t.s) in region of gonads
  • Roundworm ova from faeces (w.m)
  • Pin worm adult specimen (w.m)
  • Muscle with encysted larvae (l.s)
  • Tick adult (w.m) carrier of relapsing fever and borreliosis
  • Chicken mite (w.m) adult specimen
  • Parasitic mite of honey bee (w.m)
  • Section of diseased skin with parasites
  • Anopheles, malaria mosquito, head and mouth parts of female (w.m)
  • Common mosquito, head and mouth parts of female (w.m)
  • Bed bug (w.m) of male or female
  • Human louse (w.m) of male or female
  • Dog flea, adult male or female (w.m)
Bestellinformation: Includes 25 microscope slides with accompanying brochure.