Drucken

Mouse IgM purification kit

Lieferant: LIGATRAP TECHNOLOGIES
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

LT-145 LT-145KIT LT-145-1X1ML LT-145-1X5ML LT-145-3X1ML
LIGALT-145EA 0 EUR
LIGALT-145 LIGALT-145KIT LIGALT-145-1X1ML LIGALT-145-1X5ML LIGALT-145-3X1ML
Mouse IgM purification kit
Protein-Aufbereitungs Protein Purification Kits

LigaTrap® Mouse IgM Purification Kit with LT technology utilises novel, patented ligands, designed for IgM antibody purification. Kit provides all the necessary reagents for fast, convenient micro-scale purification of mouse IgM in just ten easy steps.


  • Designed to purify mouse IgM
  • For in vitro laboratory use
  • Binding capacity for mouse IgM is >10 mg/ml resin
  • Storage: 2 to 8 °C


LigaTrap® mouse IgM purification resin is engineered to purify high quality mouse IgM antibodies from recombinant and monoclonal sources. Resins bind an exceptionally broad range of recombinant, monoclonal and human/animal source polyclonal immunoglobulins.


Each spin column can be used, regenerated, and used up to ten times with minimal
loss in binding capacity.


Lieferumfang: Each kit contains ten microspin columns prefilled with 0,1 ml of LigaTrap® mouse IgM purification resin, buffers and collection tubes.


Achtung! Bitte beachten: Not for diagnostic or therapeutic use. Not for human or animal consumption.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR