Drucken

Human IgA purification kit, LigaTrap®

Lieferant: LIGATRAP TECHNOLOGIES
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

LT-146 LT-146KIT LT-146-1X1ML LT-146-1X5ML LT-146-3X1ML
LIGALT-146EA 0 EUR
LIGALT-146 LIGALT-146KIT LIGALT-146-1X1ML LIGALT-146-1X5ML LIGALT-146-3X1ML
Human IgA purification kit, LigaTrap®
Protein-Aufbereitungs Protein Purification Kits

LigaTrap® Human IgA Purification Kit with LT technology utilises novel, patented ligands, designed for IgA antibody purification. Kit provides all the necessary reagents for fast, convenient micro-scale purification of human IgA in just ten easy steps.


  • Designed to purify human IgA
  • For in vitro laboratory use
  • Binding capacity for monoclonal human IgA is >15 mg/ml resin
  • Storage: 2 to 8 °C


LigaTrap® Human IgA purification resin is engineered to purify high quality human IgA antibodies from recombinant and monoclonal sources. Resins bind an exceptionally broad range of recombinant, monoclonal and human/animal source polyclonal immunoglobulins.


Each spin column can be used, regenerated, and used up to ten times with minimal
loss in binding capacity.


Lieferumfang: Each kit contains ten microspin columns prefilled with 0,10 ml of LigaTrap® human IgA purification resin, buffers and collection tubes.


Achtung! Bitte beachten: Not for diagnostic or therapeutic use. Not for human or animal consumption.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR