VWR®, Blocking buffer, non-fat powdered milk
Lieferant: VWR Chemicals
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
M232-39.8G-5PKEA
181
EUR
M232-39.8G-5PK
VWR®, Blocking buffer, non-fat powdered milk
Puffer
Elektrophorese-Puffer
Blockier-Puffer
Each pack of powdered blocking buffer makes one liter of 1X solution.
- Easy to use
- Optimise assay performance using high quality blocking buffers