Drucken
Miller's LB broth, Corning®
Lieferant: MEDIATECH
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
46-050-CM
MDTC46-050-CMEA 240.31 EUR
MDTC46-050-CM
Miller's LB broth, Corning®
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Miller's LB broth is based on the original Lysogeny Broth (LB) media formulation developed by Luria and Bertani, which was designed for the rapid propagation of recombinant E. coli strains.
  • 20-fold concentration sodium chloride compared to the original formulation
  • Widely used for the growth and maintenance of cells
  • Work with plasmid DNA and recombinant protein production
Lieferumfang: Contents: Casein peptone (10 g/L), UF Yeast Powder (5 g/L), NaCl (10 g/L)