Drucken
Laboratory marker
Lieferant: MEDITE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
47-1070-00
MEDT47-1070-00EA 68.5 EUR
MEDT47-1070-00
Laboratory marker
Schreibstifte Labormarker
Marker for labelling cassettes and microscope slides with a high contrast black ink. These are water-based, therefor no early drying out is possible.
  • Water resistant, odour-free and non toxic
  • Resistant against most laboratory chemicals
  • No pretreatment of surfaces necessary