Drucken
Shelving systems, open grid, MetroMax i®
Lieferant: Metro International
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
X566EGX3
METOX566EGX3EA 2800 EUR
METOX566EGX3
Shelving systems, open grid, MetroMax i®
Möbel Regale
Corrosion proof shelves and posts with removable polymer mats.
  • Consist of four open grid shelves and posts, wedge connectors and polyurethane casters (2-swivel and 2-brake)
  • Microban® antimicrobial product protection is built into every shelf mat and touch points
  • Shelving up to 900 lbs. (408 kg) per shelf