Drucken
Masterflex® Self-Priming Petroleum Centrifugal Pumps, Avantor®
Lieferant: Avantor Fluid Handling
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
79735-11
MFLX79735-11EA 1880 EUR
MFLX79735-11
Masterflex® Self-Priming Petroleum Centrifugal Pumps, Avantor®
Pumpen Kreiselpumpen
Compatible with three distinct fuel classifications.
  • Models for the following fluid groups:- Gasoline, kerosene, avgas, and jet fuel- Biodiesel, fuel oil, and diesel- Ethanol and E85 blend
  • Available in both self-priming and standard end suction configurations
  • Custom configurations are a phone call away to our Application Specialists
These close-coupled pumps have been specifically engineered for compatibility with clean, nonabrasive petroleum products. Transfer and delivery of various fuels such as gasoline, ethanol, biodiesel, and fuel oils are just a few of the petroleum-based products these pumps can handle.