Drucken
Plastic Tube Brushes
Lieferant: MICRONOVA
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
BRPL-1218 BRPL-3418 BRPL-118 BRPL-1418 BRPL-25S36
MICRBRPL-1218EA 57.5 EUR
MICRBRPL-1218 MICRBRPL-3418 MICRBRPL-118 MICRBRPL-1418 MICRBRPL-25S36
Plastic Tube Brushes
Bürsten
These durable, 100% polypropylene brushes clean finely polished stainless steel parts without scratching.
  • 100% polypropylene bristles and handle
  • Thin core and handles are flexible enough to bend through fittings
  • Available in a range of bristle diameters and handle lengths
Brushes feature narrow center cores that are flexible enough to pass through elbow tank fittings.
Zertifizierungen: FDA compliant materials.