Drucken
Detergents for glassware washers, ProCare Lab 11 MA
Lieferant: MIELE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
10266720
MIEL10266720EA 118 EUR
MIEL10266720
Detergents for glassware washers, ProCare Lab 11 MA
Reinigungsmittel Waschmittel und Oberflächenreiniger
Powder, mildly alkaline, foam-retardant action detergent for machine based cleaning of laboratory glassware and laboratory utensils.
  • Provides good protection to surfaces
  • Removes blood, protein and food residues
  • Free from oxidative agents
ProCare Lab 11 MA is for general-purpose use in medical, biological and chemical laboratories.
Note: Suitable for stainless steel, laboratory glassware, ceramics, plastics such as PE, PP, PVDF, PTFE. Conditionally suitable for use on non ferrous metals and aluminium. Not suitable for anodised aluminium.