VWR®, Taq DNA polymerase, 500 U
Lieferant: VWR ChemicalsGesamtbewertung: 0
Bewertung: von 5
Taq DNA Polymerase is ideal for general PCR and primer extension reactions to amplify DNA templates with high sensitivity. PCR can be initiated from as few as 6 primed templates. Simply add 0,5 µl of Taq DNA Polymerase to a 25 µl PCR reaction.
- Highly purified, DNA-free, and free of bacterial genomic DNA
- Amplify DNA templates with high sensitivity
- Amplify single-copy and low abundance templates
One unit of activity incorporates 10 nmoles of dNTP into acid-insoluble DNA in 30 minutes at 72 °C.
The PCR kit with Taq DNA Polymerase includes separate solutions of all material needed for a PCR reaction, including dNTPs, reaction buffer with and without MgCl2, 25 mM MgCl2, Taq Polymerase, and sterile water.
Lieferumfang: Contains Taq DNA Polymerase, 500 U (5 U/µl) and Taq Buffer, 10X, 1 ml. Sufficient for 200× 25 µl amplifications.