Drucken
Chamber slides, C-Slide
Lieferant: NANOENTEK
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
CS-050
NANOCS-050EA 134 EUR
NANOCS-050
Chamber slides, C-Slide
Objektträger Kammerobjektträger
Cell counting chamber slides for use with automated cell counter (Countess and Countess II).
  • No need of coverslips
  • Eliminates the need of washing and reuse
  • Reduced exposures to the infectious samples and contamination
  • Guaranteed reproducibility and reliability
Made of sturdy and strong quartz grade optical plastic.