Drucken

Anti-MTMR14 Mouse Monoclonal Antibody [clone: 6F8]

Lieferant: OriGene
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

CF501607
ORIGCF501607EA 833 EUR
ORIGCF501607
Anti-MTMR14 Mouse Monoclonal Antibody [clone: 6F8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MTMR14
Clonality: Monoclonal
Clone: 6F8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR