Drucken

Anti-CBWD1 Mouse Monoclonal Antibody [clone: 4D4]

Lieferant: OriGene
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

CF501609
ORIGCF501609EA 833 EUR
ORIGCF501609
Anti-CBWD1 Mouse Monoclonal Antibody [clone: 4D4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CBWD1
Clonality: Monoclonal
Clone: 4D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR