Drucken

Anti-EGFR Mouse Monoclonal Antibody [clone: 1E5]

Lieferant: OriGene
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

CF801938
ORIGCF801938EA 833 EUR
ORIGCF801938
Anti-EGFR Mouse Monoclonal Antibody [clone: 1E5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR