Drucken

Anti-DDX39A Rabbit Polyclonal Antibody

Lieferant: OriGene
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

TA308418
ORIGTA308418EA 868 EUR
ORIGTA308418
Anti-DDX39A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: DDX39A (DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR