Drucken

Anti-GPR183 Rabbit Polyclonal Antibody

Lieferant: OriGene
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

TA316825
ORIGTA316825EA 715 EUR
ORIGTA316825
Anti-GPR183 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: GPR183 (G protein-coupled receptor 183)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR