Drucken

Anti-NPM1 Rabbit Polyclonal Antibody

Lieferant: OriGene
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

TA590345
ORIGTA590345EA 669 EUR
ORIGTA590345
Anti-NPM1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:5000-1:20000; ELISA: 1:100-1:2000; IHC: 1:10-1:2000; IHC-P 1:250-1:2000

Type: Primary
Antigen: NPM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR