Drucken

Anti-PTGS2 Mouse Monoclonal Antibody [clone: 4C4]

Lieferant: OriGene
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

TA805224
ORIGTA805224EA 621 EUR
ORIGTA805224
Anti-PTGS2 Mouse Monoclonal Antibody [clone: 4C4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PTGS2
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR