Transduction enhancer, AdenoMag™
Lieferant: OZ BIOSCIENCESGesamtbewertung: 0
Bewertung: von 5
AdenoMag™ transfection reagent is a magnetic nanoparticles formulation specifically dedicated to enhance Adenovirus and Adeno Associated Virus (AAV) infection and transduction capacities. Based on Magnetofection™ technology, it allows concentrating the complete applied dose of viral particles onto cells within minutes, inducing a significant improvement of virus infectivity with extremely low vector doses.
- Increase transduction efficiency
- Accelerate the transduction process
- Efficient for hard-to-infect and non permissive cells
- Concentrate the viral dose onto the cells
- Synchronise cells adsorption/infection without modifications of the viruses
AdenoMag™ reagent demonstrates an exceptionally high efficiency to promote, control and assist Adenoviral and AAV transductions in such a way that no molecular biology processes or biochemical modifications are required.
Store at +4 °C.
Bestellinformation: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
Lieferumfang: Shipped at room temperature.