Drucken
DNA transfection reagent, DreamFect™
Lieferant: OZ BIOSCIENCES
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
DF40500 DF41000 DF45000
OZBIDF40500EA 171 EUR
OZBIDF40500 OZBIDF41000 OZBIDF45000
DNA transfection reagent, DreamFect™
Transfektionsreagenzien
DreamFect™ transfection reagent is based on an innovative biochemical method. It contains both a lipidic part and a cationic part allowing the association of nucleic acids with this transfection reagent and their delivery inside cells. Due to its unique properties, it releases large amounts of DNA in the cytosol increasing DNA nuclear delivery (Tee-technology). Consequently, it is highly efficient at low doses of nucleic acids and less cationic lipids are used so potential secondary effects are avoided.
  • Exceptional efficiency
  • High reproducibility
  • Broad range of cell types
  • Non-toxic
  • Serum compatible
  • Simple, rapid, and easy to use
Store at +4 °C.
Lieferumfang: Shipped at room temperature.