DNA transfection reagent, FlyFectIN™
Lieferant: OZ BIOSCIENCESGesamtbewertung: 0
Bewertung: von 5
FlyFectIN™ transfection reagent is a powerful reagent based on Tee-technology and specifically designed to obtain highly efficient and reproducible transfection of insect cells. It is adapted to the delivery of all types of nucleic acids. It can be used for many applications including the production of recombinant protein using Baculovirus expression system.
- Dedicated to insect cells
- Ideal for production of recombinant protein using Baculovirus expression system
- Non toxic
- Serum compatible
- Simple, ready to use and rapid
FlyFectIN™ has been effectively used with several insect cells. Store at 4 °C.
Lieferumfang: Shipped at room temperature.