Drucken
DNA transfection reagent, FlyFectIN™
Lieferant: OZ BIOSCIENCES
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
FF50500 FF51000 FF55000
OZBIFF50500EA 162.95 EUR
OZBIFF50500 OZBIFF51000 OZBIFF55000
DNA transfection reagent, FlyFectIN™
Transfektionsreagenzien
FlyFectIN™ transfection reagent is a powerful reagent based on Tee-technology and specifically designed to obtain highly efficient and reproducible transfection of insect cells. It is adapted to the delivery of all types of nucleic acids. It can be used for many applications including the production of recombinant protein using Baculovirus expression system.
  • Dedicated to insect cells
  • Ideal for production of recombinant protein using Baculovirus expression system
  • Non toxic
  • Serum compatible
  • Simple, ready to use and rapid
FlyFectIN™ has been effectively used with several insect cells. Store at 4 °C.
Lieferumfang: Shipped at room temperature.