Drucken
Transfection reagents, FluoMag™
Lieferant: OZ BIOSCIENCES
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
FN10100 FC10100 FP10100 FS10100 FV10100
OZBIFN10100EA 134.38 EUR
OZBIFN10100 OZBIFC10100 OZBIFP10100 OZBIFS10100 OZBIFV10100
Transfection reagents, FluoMag™
Transfektionsreagenzien
FluoMag™ reagents are fluorescently-labeled magnetic nanoparticles for Magnetofection™ applications. FluoMag™ transfection reagents are as efficient as their non labelled counterparts and allow visualisation of the nanoparticles in vitro during experiments.
  • Advantages of Magnetofection™ technology
  • Easy cell labelling and tracking
  • Non toxic
  • Serum compatible
Store at +4 °C and protect from the light.
Bestellinformation: This reagent needs to be used with a magnetic plate, available separately.
Lieferumfang: Shipped at room temperature.