Transfection reagents, FluoMag™
Lieferant: OZ BIOSCIENCESGesamtbewertung: 0
Bewertung: von 5
FluoMag™ reagents are fluorescently-labeled magnetic nanoparticles for Magnetofection™ applications. FluoMag™ transfection reagents are as efficient as their non labelled counterparts and allow visualisation of the nanoparticles in vitro during experiments.
- Advantages of Magnetofection™ technology
- Easy cell labelling and tracking
- Non toxic
- Serum compatible
Store at +4 °C and protect from the light.
Bestellinformation: This reagent needs to be used with a magnetic plate, available separately.
Lieferumfang: Shipped at room temperature.