Drucken

DNA transfection reagent, HYPE-5™

Lieferant: OZ BIOSCIENCES
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

HY01500 HY03000 HY15000 HY30000 HYR10003 HYR10015 HYR20030
OZBIHY01500EA 222.59 EUR
OZBIHY01500 OZBIHY03000 OZBIHY15000 OZBIHY30000 OZBIHYR10003 OZBIHYR10015 OZBIHYR20030
DNA transfection reagent, HYPE-5™
Transfektionsreagenzien

HYPE-5™ transfection kit is dedicated to achieve high yield protein expression in mammalian cells. The kit has been designed for maximum recombinant protein expression in HEK293 and CHO cells grown in suspension. It is composed of the HYPE-5™ transfection reagent (a lipofection reagent) and HYPE-5™ Blast reagent (improves protein production in CHO cells). The HYPE-5™ transfection kit is ideal for bioreactor, spinner or flasks.


  • High yield recombinant protein production
  • Suitable for both HEK293 and CHO cells growing in suspension
  • Compatible with any synthetic or regular media used for protein production
  • Completely animal origin free


Store HYPE-5™ reagent at –20 °C and HYPE-5™ Blast at +4 °C.


Lieferumfang: Shipped at room temperature.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR